2,4-Dichloro-6-methylbenzoic acid 96%

Supplier: Ambeed
Warning

A788336-100MG A788336-250MG
77171-290EA 161.71 USD
77171-290 77171-292
2,4-Dichloro-6-methylbenzoic acid 96%
2,4-Dichloro-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MDL Number: MFCD06208457
CAS Number: 65410-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR