6,7-Dihydro-[1,4]dioxino[2',3':4,5]benzo[1,2-d]thiazol-2-amine 97%

Supplier: Ambeed
Warning

A787019-100MG A787019-1G A787019-250MG
77199-484EA 50 USD
77199-484 77199-486 77199-488
6,7-Dihydro-[1,4]dioxino[2',3':4,5]benzo[1,2-d]thiazol-2-amine 97%
6,7-Dihydro-[1,4]dioxino[2',3':4,5]benzo[1,2-d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MDL Number: MFCD00560023
CAS Number: 313223-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR