1-(4-Chlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride 97%

Supplier: Ambeed
Warning

A784316-1G A784316-250MG A784316-5G
77199-474EA 42 USD
77199-474 77199-476 77199-478
1-(4-Chlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride 97%
1-(4-Chlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO·ClH
MDL Number: MFCD00084951
CAS Number: 1798-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR