5-Tetradecyldihydrofuran-2(3H)-one 95%

Supplier: Ambeed
Warning

A783535-100MG A783535-250MG
77171-272EA 87.55 USD
77171-272 77569-478
5-Tetradecyldihydrofuran-2(3H)-one 95%
5-Tetradecyldihydrofuran-2(3H)-one

Precaución: For research use only.

Formula: C₁₈H₃₄O₂
MW: 282.46 g/mol
MDL Number: MFCD01725883
CAS Number: 502-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR