4,5-Dihydrofuran-3-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4,5-Dihydrofuran-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A783472-100MG A783472-1G A783472-250MG A783472-50MG A783472-5G
77171-264EA 42.99 USD
77171-264 77171-266 77171-268 77171-270 77893-541
4,5-Dihydrofuran-3-boronic acid, pinacol ester 98%
4,5-Dihydrofuran-3-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₇BO₃
MDL Number: MFCD13182120
CAS Number: 1046812-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR