1-Boc-4-(4-piperidinyl)-piperazine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(piperidin-4-yl)piperazine-1-carboxylate

A783118-100MG A783118-1G A783118-250MG A783118-25G A783118-5G
77199-462EA 12 USD
77199-462 77199-464 77199-466 77199-468 77552-426
1-Boc-4-(4-piperidinyl)-piperazine 95%
1-Boc-4-(4-piperidinyl)-piperazine

Precaución: for Research Use Only

Formula: C₁₄H₂₇N₃O₂
MW: 269.383 g/mol
MDL Number: MFCD04974009
CAS Number: 205059-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR