2-Fluoro-p-toluidine 98%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoro-4-methylaniline, 4-Amino-3-fluorotoluene

A782821-10G A782821-1G A782821-25G A782821-5G
77199-454EA 8 USD
77199-454 77199-456 77199-458 77199-460
2-Fluoro-p-toluidine 98%
2-Fluoro-p-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
MDL Number: MFCD00040975
CAS Number: 452-80-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR