N-(2-Acetamido)iminodiacetic acid, monosodium salt 96%

Supplier: Ambeed
Warning

Synonyms: Sodium 2-((2-amino-2-oxoethyl)(carboxymethyl)amino)acetate, ADA monosodium salt

A782717-100G A782717-25G A782717-500G
77171-258EA 40 USD
77171-258 77171-260 77171-262
N-(2-Acetamido)iminodiacetic acid, monosodium salt 96%
N-(2-Acetamido)iminodiacetic acid, monosodium salt

Precaución: for Research Use Only

Formula: C₆H₉N₂O₅·Na
MW: 212.136 g/mol
MDL Number: MFCD00065483
CAS Number: 7415-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR