Ethyl 6-chloro-[1,2,4]triazolo[4,3-b]pyridazine-3-carboxylate 95%

Supplier: Ambeed
Warning

A781938-100MG A781938-1G A781938-250MG A781938-5G
77171-248EA 87 USD
77171-248 77171-250 77171-252 77171-254
Ethyl 6-chloro-[1,2,4]triazolo[4,3-b]pyridazine-3-carboxylate 95%
Ethyl 6-chloro-[1,2,4]triazolo[4,3-b]pyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇ClN₄O₂
MDL Number: MFCD11868226
CAS Number: 75680-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR