Pyrazolo[1,5-a]pyridine-7-carboxylic acid 97%

Supplier: Ambeed
Warning

A781935-100MG A781935-1G A781935-250MG
77171-242EA 56.65 USD
77171-242 77171-244 77171-246
Pyrazolo[1,5-a]pyridine-7-carboxylic acid 97%
Pyrazolo[1,5-a]pyridine-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
MDL Number: MFCD06739030
CAS Number: 474432-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR