2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-4-carboxylic acid

A779207-1G A779207-5G
77199-438EA 21.63 USD
77199-438 77199-440
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid 98%
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₄
MW: 202.12 g/mol
MDL Number: MFCD01631473
CAS Number: 126120-85-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR