4-Chloropicolinohydrazide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-(hydrazinecarbonyl)pyridine

A778943-250MG A778943-5G
77171-226EA 32.96 USD
77171-226 77171-228
4-Chloropicolinohydrazide 98%
4-Chloropicolinohydrazide

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O
MW: 171.5843 g/mol
MDL Number: MFCD01704394
CAS Number: 73771-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR