6-Chloro-[1,1'-biphenyl]-3-amine 95%

Supplier: Ambeed
Warning

A778083-100MG A778083-250MG
77171-224EA 88 USD
77171-224 77893-538
6-Chloro-[1,1'-biphenyl]-3-amine 95%
6-Chloro-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClN
MDL Number: MFCD12913724
CAS Number: 56970-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR