Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate 96%

Supplier: Ambeed
Warning

A777369-1G A777369-250MG
77171-220EA 23.69 USD
77171-220 77171-222
Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate 96%
Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD02186220
CAS Number: 67625-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR