GSK-J1 (Free acid) 98%

Supplier: Ambeed
Warning

Synonyms: 3-{[2-(pyridin-2-yl)-6-(2,3,4,5-tetrahydro-1H-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoic acid, N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine, 3-((2-(Pyridin-2-yl)-6-(1,2,4,5-tetrahydro-3H-benzo[d]azepin-3-yl)pyrimidin-4-yl)amino)propanoic acid, GSK-J1

A773094-10MG A773094-1MG A773094-50MG A773094-5MG
77171-200EA 64.89 USD
77171-200 77171-202 77171-204 77171-206
GSK-J1 (Free acid) 98%
GSK-J1 (Free acid)

Precaución: for Research Use Only

Formula: C₂₂H₂₃N₅O₂
MW: 398.45 g/mol
MDL Number: MFCD22683851
CAS Number: 1373422-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR