9-Methyl-3-methylene-2,3-dihydro-1H-carbazol-4(9H)-one 95+%

Supplier: Ambeed
Danger

A772796-1G A772796-25G A772796-5G
77199-410EA 24.72 USD
77199-410 77199-412 77199-414
9-Methyl-3-methylene-2,3-dihydro-1H-carbazol-4(9H)-one 95+%
9-Methyl-3-methylene-2,3-dihydro-1H-carbazol-4(9H)-one

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO
MW: 211.25 g/mol
MDL Number: MFCD08063682
CAS Number: 99614-64-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR