4-(N-(4-Cyclohexylbenzyl)-2-((2,3,4,5,6-pentafluoro-N-methylphenyl)sulfonamido)acetamido)-2-hydroxybenzoic acid 99+%

Supplier: Ambeed
Warning

Synonyms: BP-1-102

A772321-10MG A772321-1MG A772321-25MG A772321-2MG A772321-5MG
77199-400EA 169.95 USD
77199-400 77199-402 77199-404 77199-406 77199-408
4-(N-(4-Cyclohexylbenzyl)-2-((2,3,4,5,6-pentafluoro-N-methylphenyl)sulfonamido)acetamido)-2-hydroxybenzoic acid 99+%
4-(N-(4-Cyclohexylbenzyl)-2-((2,3,4,5,6-pentafluoro-N-methylphenyl)sulfonamido)acetamido)-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₂₉H₂₇F₅N₂O₆S
MW: 626.59 g/mol
MDL Number: MFCD29477477
CAS Number: 1334493-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR