1-(2H-Pyrazol-3-yl)ethanone hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-1H-pyrazole hydrochloride, 1-(1H-Pyrazol-3-yl)ethanone hydrochloride

A771751-100MG A771751-250MG A771751-1G
77199-390EA 23.69 USD
77199-390 77199-392 77747-522
1-(2H-Pyrazol-3-yl)ethanone hydrochloride 95%
1-(2H-Pyrazol-3-yl)ethanone hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇ClN₂O
MW: 146.58 g/mol
MDL Number: MFCD00204160
CAS Number: 175277-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR