4-Bromo-1H-benzo[d]imidazol-2(3H)-one 96%
Supplier: Ambeed
|
Warning
|
A770726-100MG
A770726-1G
77199-386EA
116.39
USD
77199-386
77199-388
4-Bromo-1H-benzo[d]imidazol-2(3H)-one 96%
4-Bromo-1H-benzo[d]imidazol-2(3H)-one
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂O MW: 213.03 g/mol |
MDL Number:
MFCD17267704 CAS Number: 40644-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...