2,4-Dichloro-7,8-dihydroquinolin-5(6H)-one 97%

Supplier: Ambeed
Warning

A770000-100MG A770000-250MG
77199-376EA 154.5 USD
77199-376 77199-378
2,4-Dichloro-7,8-dihydroquinolin-5(6H)-one 97%
2,4-Dichloro-7,8-dihydroquinolin-5(6H)-one

Precaución: for Research Use Only

Formula: C₉H₇Cl₂NO
MW: 216.06 g/mol
MDL Number: MFCD18253894
CAS Number: 1028330-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR