[4-(methoxycarbonyl)-3-methylphenyl]boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Methoxycarbonyl)-3-methylphenyl)boronic acid, 3-Methyl-4-(methoxycarbonyl)phenylboronic acid

A769603-250MG A769603-100MG A769603-5G
77171-182EA 64 USD
77171-182 77893-521 77893-522
[4-(methoxycarbonyl)-3-methylphenyl]boronic acid 98%
[4-(methoxycarbonyl)-3-methylphenyl]boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
MDL Number: MFCD18432535
CAS Number: 603122-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR