N-(4-Bromo-3,5-dimethylphenyl)-2,2,2-trifluoroacetamide 97%

Supplier: Ambeed
Warning

A766911-250MG A766911-5G
77199-362EA 32.96 USD
77199-362 77199-364
N-(4-Bromo-3,5-dimethylphenyl)-2,2,2-trifluoroacetamide 97%
N-(4-Bromo-3,5-dimethylphenyl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₃NO
MW: 296.08 g/mol
MDL Number: MFCD27949080
CAS Number: 306297-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR