Ethyl 2,4,5,6-tetrahydrocyclopenta[c]pyrrole-1-carboxylate 97%

Supplier: Ambeed
Warning

A766234-1G A766234-250MG
77199-358EA 307.97 USD
77199-358 77199-360
Ethyl 2,4,5,6-tetrahydrocyclopenta[c]pyrrole-1-carboxylate 97%
Ethyl 2,4,5,6-tetrahydrocyclopenta[c]pyrrole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.21 g/mol
MDL Number: MFCD12924739
CAS Number: 128812-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR