(R)-3,3'-Bis(3,5-di-tert-butylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%

Supplier: Ambeed
Warning

A765091-100MG A765091-250MG A765091-50MG
77199-340EA 73 USD
77199-340 77199-342 77199-344
(R)-3,3'-Bis(3,5-di-tert-butylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%
(R)-3,3'-Bis(3,5-di-tert-butylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₄₈H₅₃O₄P
MDL Number: MFCD28139326
CAS Number: 861909-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR