2,3-Bis((tert-butoxycarbonyl)amino)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-Dap(Boc)-OH

A764425-1G A764425-250MG
77171-158EA 91.67 USD
77171-158 77171-160
2,3-Bis((tert-butoxycarbonyl)amino)propanoic acid 95%
2,3-Bis((tert-butoxycarbonyl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₆
MW: 304.34 g/mol
MDL Number: MFCD16036256
CAS Number: 104010-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR