(2,4-Dibromophenyl)hydrazine hydrochloride 96%

Supplier: Ambeed
Warning

A764258-10G A764258-1G A764258-250MG A764258-5G
77199-320EA 153.47 USD
77199-320 77199-322 77199-324 77199-326
(2,4-Dibromophenyl)hydrazine hydrochloride 96%
(2,4-Dibromophenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇Br₂ClN₂
MW: 302.39 g/mol
MDL Number: MFCD04966814
CAS Number: 149998-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR