3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 3-((4-Methylpiperazin-1-yl)methyl)benzonitrile

A763164-1G A763164-250MG A763164-5G
77171-152EA 30.9 USD
77171-152 77171-154 77171-156
3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile 95%
3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₃
MW: 215.294 g/mol
MDL Number: MFCD07772861
CAS Number: 859850-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR