1,3-Di(adamantan-1-yl)-4,5-dihydro-1H-imidazol-3-ium chloride 98%
Supplier: Ambeed
|
Warning
|
A762444-250MG
A762444-100MG
A762444-1G
77199-308EA
41.2
USD
77199-308
77618-102
77747-507
1,3-Di(adamantan-1-yl)-4,5-dihydro-1H-imidazol-3-ium chloride 98%
1,3-Di(adamantan-1-yl)-4,5-dihydro-1H-imidazol-3-ium chloride
Precaución: for Research Use Only
Formula:
C₂₃H₃₅ClN₂ MW: 374.99 g/mol |
MDL Number:
MFCD17013973 CAS Number: 871126-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...