6-Bromo-5-methylimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A759791-100MG A759791-250MG
77199-296EA 39.14 USD
77199-296 77199-298
6-Bromo-5-methylimidazo[1,2-a]pyridine 97%
6-Bromo-5-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.05 g/mol
MDL Number: MFCD20489149
CAS Number: 116355-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR