Castanospermine 98%

Supplier: Ambeed
Warning

Synonyms: (1S,6S,7R,8R,8aR)-Octahydroindolizine-1,6,7,8-tetraol

A758766-100MG A758766-250MG A758766-50MG
77199-272EA 224.54 USD
77199-272 77199-274 77199-276
Castanospermine 98%
Castanospermine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
MDL Number: MFCD00017555
CAS Number: 79831-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR