6-Methoxy-1H-indazole-5-carboxylic acid 96%

Supplier: Ambeed
Warning

A758424-100MG A758424-1G A758424-250MG
77171-122EA 175.1 USD
77171-122 77171-124 77171-126
6-Methoxy-1H-indazole-5-carboxylic acid 96%
6-Methoxy-1H-indazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MW: 192.17 g/mol
MDL Number: MFCD11007888
CAS Number: 1082041-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR