(2,6-Bis(benzyloxy)pyridin-3-yl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A757865-250MG
A757865-5G
A757865-1G
77199-262EA
21
USD
77199-262
77573-442
77747-493
(2,6-Bis(benzyloxy)pyridin-3-yl)boronic acid 98%
(2,6-Bis(benzyloxy)pyridin-3-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₉H₁₈BNO₄ |
MDL Number:
MFCD18434424 CAS Number: 2096339-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...