Methyl 4-amino-3-bromo-5-methylbenzoate 97%

Supplier: Ambeed
Warning

A757725-1G A757725-250MG A757725-5G
77171-108EA 23 USD
77171-108 77171-110 77893-497
Methyl 4-amino-3-bromo-5-methylbenzoate 97%
Methyl 4-amino-3-bromo-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MDL Number: MFCD07021337
CAS Number: 900019-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR