1-Bromo-8-iodonaphthalene 98%

Supplier: Ambeed
Warning

A757206-1G A757206-250MG A757206-5G A757206-25G
77171-102EA 47 USD
77171-102 77171-104 77171-106 77893-496
1-Bromo-8-iodonaphthalene 98%
1-Bromo-8-iodonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆BrI
MDL Number: MFCD11656276
CAS Number: 4044-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR