(S)-3-Amino-butyric acid methyl ester hydrochloride 96%

Supplier: Ambeed
Warning

A756729-250MG A756729-5G A756729-1G A756729-25G
77171-092EA 23 USD
77171-092 77171-094 77614-338 77893-495
(S)-3-Amino-butyric acid methyl ester hydrochloride 96%
(S)-3-Amino-butyric acid methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂·ClH
MDL Number: MFCD12068617
CAS Number: 139243-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR