1,2-Ethanedisulfonic acid disodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium ethane-1,2-disulphonate

A756609-100G A756609-25G A756609-5G A756609-1G
77199-250EA 76 USD
77199-250 77199-252 77199-254 77747-488
1,2-Ethanedisulfonic acid disodium salt 98%
1,2-Ethanedisulphonic acid disodium salt

Precaución: for Research Use Only

Formula: C₂H₄Na₂O₆S₂
MDL Number: MFCD00024998
CAS Number: 5325-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR