(3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate 95%

Supplier: Ambeed
Warning

A756519-100G A756519-25G
77199-246EA 202 USD
77199-246 77199-248
(3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate 95%
(3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₁₁
MDL Number: MFCD00069794
CAS Number: 25878-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR