2-Boc-6-hydroxy-2-azaspiro[3.4]octane 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-2-propanyl 6-hydroxy-2-azaspiro[3.4]octane-2-carboxylate, tert-Butyl 6-hydroxy-2-azaspiro[3.4]octane-2-carboxylate

A755173-100MG A755173-250MG A755173-5G A755173-1G
77171-088EA 35.02 USD
77171-088 77171-090 77573-432 77614-328
2-Boc-6-hydroxy-2-azaspiro[3.4]octane 98%
2-Boc-6-hydroxy-2-azaspiro[3.4]octane

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.3 g/mol
MDL Number: MFCD22415223
CAS Number: 1363381-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR