1-Bromo-4-chloro-3-fluoro-2-methoxybenzene 97%

Supplier: Ambeed
Warning

A754712-1G A754712-5G
77199-240EA 376.98 USD
77199-240 77573-428
1-Bromo-4-chloro-3-fluoro-2-methoxybenzene 97%
1-Bromo-4-chloro-3-fluoro-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.46 g/mol
MDL Number: MFCD22414766
CAS Number: 1414870-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR