2-Fluoro-4-methylbenzeneboronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

A753232-100MG A753232-1G A753232-250MG A753232-5G
77171-080EA 19 USD
77171-080 77171-082 77171-084 77171-086
2-Fluoro-4-methylbenzeneboronic acid pinacol ester ≥97%
2-Fluoro-4-methylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂
MDL Number: MFCD18779806
CAS Number: 1165936-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR