1-(4-Chloro-2-methoxyphenyl)ethanone 97%

Supplier: Ambeed
Warning

A753228-1G A753228-250MG
77199-236EA 90.64 USD
77199-236 77199-238
1-(4-Chloro-2-methoxyphenyl)ethanone 97%
1-(4-Chloro-2-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.61 g/mol
MDL Number: MFCD11553429
CAS Number: 60207-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR