Ethyl 4-amino-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A752141-25G A752141-5G
77199-232EA 47.38 USD
77199-232 77199-234
Ethyl 4-amino-2-methylbenzoate 98%
Ethyl 4-amino-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD08458830
CAS Number: 74450-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR