2-Bromo-6-fluoro-3-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A752123-100MG A752123-250MG
77171-076EA 221.45 USD
77171-076 77171-078
2-Bromo-6-fluoro-3-nitrobenzoic acid 95%
2-Bromo-6-fluoro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrFNO₄
MW: 264.01 g/mol
MDL Number: MFCD22581538
CAS Number: 1036388-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR