4-Aminophenyl 4-Aminobenzoate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Aminobenzoic acid 4-Aminophenyl ester, 4-Aminophenyl 4-aminobenzoate
A751901-100MG
A751901-1G
A751901-250MG
77171-070EA
6
USD
77171-070
77171-072
77171-074
4-Aminophenyl 4-Aminobenzoate 97%
4-Aminophenyl 4-Aminobenzoate
Precaución: for Research Use Only
Formula:
C₁₃H₁₂N₂O₂ MW: 228.25 g/mol |
MDL Number:
MFCD16038468 CAS Number: 20610-77-9 UN: 2811 ADR: 6.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...