1,11-Dibromoundecane 96%

Supplier: Ambeed
Warning

Synonyms: Undecamethylene dibromide

A751525-100G A751525-1G A751525-5G
77199-226EA 332.69 USD
77199-226 77199-228 77199-230
1,11-Dibromoundecane 96%
1,11-Dibromoundecane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₉CH₂Br
MW: 314.1 g/mol
MDL Number: MFCD00000224
CAS Number: 16696-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR