1-Acetylpyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Pyrrolidin-1-yl)ethanone

A751159-1G A751159-25G A751159-5G
77171-042EA 31.93 USD
77171-042 77171-044 77171-046
1-Acetylpyrrolidine 98%
1-Acetylpyrrolidine

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MW: 113.16 g/mol
MDL Number: MFCD00014096
CAS Number: 4030-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR