7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene 98+%

Supplier: Ambeed
Warning

A750648-1G A750648-250MG A750648-5G
77171-036EA 19 USD
77171-036 77171-038 77171-040
7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene 98+%
7-Bromo-2-(chloroacetyl)-9,9-difluorofluorene

Precaución: for Research Use Only

Formula: C₁₅H₈BrClF₂O
MDL Number: MFCD28400046
CAS Number: 1378387-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR