1,2-Difluoro-3-iodobenzene 98%

Supplier: Ambeed
Warning

A749827-10G A749827-1G A749827-25G A749827-5G
77199-208EA 112.27 USD
77199-208 77199-210 77199-212 77199-214
1,2-Difluoro-3-iodobenzene 98%
1,2-Difluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂I
MW: 239.9893 g/mol
MDL Number: MFCD08458090
CAS Number: 64248-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR