(S)-1-(m-Tolyl)ethanamine 95%

Supplier: Ambeed
Warning

A748619-100MG A748619-250MG
77199-200EA 98 USD
77199-200 77199-202
(S)-1-(m-Tolyl)ethanamine 95%
(S)-1-(m-Tolyl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MDL Number: MFCD06761971
CAS Number: 138457-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR