(1-Methyl-2-nitro-1H-imidazol-5-yl)methanol 97%

Supplier: Ambeed
Warning

A746979-100MG A746979-1G A746979-250MG A746979-5G
77171-020EA 52 USD
77171-020 77171-022 77171-024 77893-476
(1-Methyl-2-nitro-1H-imidazol-5-yl)methanol 97%
(1-Methyl-2-nitro-1H-imidazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₇N₃O₃
MDL Number: MFCD06738747
CAS Number: 39070-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR